Description
- Stretchable seamless liner for great fit and dexterity
- Dots on palm and fingers for enhanced grip properties
- Good resistance to abrasion and small blade cut hazards
- Lint free glove
Specifications
Specifications
Certifications/Compliance (Europe) | CE Number, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Category II, REACH Compliant |
Chlorinated | No |
Coated Area | Palm |
Coating Material | PVC Dots |
Color | White |
Exterior Finish | Partially Textured |
Hand | Specific |
Latex Free | Yes |
Material | PVC |
Product Type | PVC-coated Industrial Glove |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
Ansell Picolon Confort Gloves >