missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI(5)P Lipid Kinase Substrate
The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A)
€ 980.00 - € 7290.00
Specifications
Content And Storage | Phosphatidylinositol 5-phosphate (PI(5)P) in 30 mM HEPES pH 7.5 buffer. |
---|---|
Form | Powder |
Product Line | Invitrogen™ |
Substrate | Phosphatidylinositol 5-phosphate (PI(5)P) |
Substrate Type | Kinase Substrate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17198825
|
Invitrogen™
A33593 |
400 nmol |
€ 980.00
400nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17285966
|
Invitrogen™
A33596 |
5000 nmol |
€ 7290.00
5000nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The PI(5)P Lipid Kinase Substrate has been specifically optimized for use with Type II PIP kinases (e.g., PIP4K2A)
- The substrate consists of solubilized phosphatidylinositol 5-phosphate (PI(5)P) in an aqueous buffer, and is used in combination with our Adapta™ Universal Kinase Assay to help identify unique kinase inhibitors.
- Validated for use with Thermo Fisher Scientific lipid kinases • Available in multiple pack sizes for convenient use
- Also available in our SelectScreen™ Kinase Profiling Service
Specifications
Phosphatidylinositol 5-phosphate (PI(5)P) in 30 mM HEPES pH 7.5 buffer. | |
Invitrogen™ | |
Kinase Substrate |
Powder | |
Phosphatidylinositol 5-phosphate (PI(5)P) | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ PI(5)P Lipid Kinase Substrate