Learn More
Phosphorus Standard Metal Solution 10,000ppm, for ICP, Aqueous Solution, Fisher Chemical
Click to view available options
Quantity:
100 mL
Packaging:
HDPE plastic bottle
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
MDL Number | 133771 |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Phosphorus solution 10 000 ppm, aqueous standard element solution