missing translation for 'onlineSavingsMsg'
Learn More

Phospho-RPS6KA1 (Ser380) Rabbit anti-Human, Mouse, CoraLite 488, Clone: 7F23, Proteintech

Rabbit Recombinant Antibody

Brand:  Proteintech CL488-80108-100UL

Product Code. 17649886

  • € 450.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Phospho-RPS6KA1 (Ser380) antibody apecifically detetects Phospho-RPS6KA1 (Ser380) antigen in Human, Mouse Samples. It is validated for Flow Cytometry.
Specifications

Specifications

Phospho-RPS6KA1 (Ser380)
7F23
CoraLite 488
BC014966
Rabbit
Protein A purification
Primary
Human, Mouse
Liquid
Flow Cytometry
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
RPS6KA1
Peptide
100 μL
6195
Store at -20°C. Avoid exposure to light.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.