missing translation for 'onlineSavingsMsg'
Learn More

Phospho-p70(S6K) (Thr229) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 135.00 - € 360.00

Specifications

Antigen Phospho-p70(S6K) (Thr229)
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17561386
View Documents
Proteintech
28988-1-AP-20UL
20 μL
€ 135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17551386
View Documents
Proteintech
28988-1-AP-100UL
100 μL
€ 360.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Phospho-p70(S6K) (Thr229) antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human Phospho-p70(S6K) (Thr229). This antibody recognizes human antigen. The Phospho-p70(S6K) (Thr229) antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

Phospho-p70(S6K) (Thr229)
Polyclonal
Rabbit
6198
Primary
Western Blot, ELISA
Unconjugated
Human
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.