Learn More
Hanna Instruments™ pHep™ Instruments pH Tester
Provides non-technical personnel with a simple, inexpensive solution to accurate pH measurement. Hanna Instruments™ pHep™ Instruments pH Tester includes a 2cm long junction that can be pulled out to expose a fresh section to renew the pHep's life.
Brand:  Hanna Instruments™ HI98107
Product Code. 10110741
  • € 132.00 / Each
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Description
Provides non-technical personnel with a simple, inexpensive solution to accurate pH measurement.
  • Extractable Junction
  • ±0.1 pH Accuracy
  • Two-Point Calibration
  • 2cm long junction that can be pulled out to expose a fresh section to effectively renew the pHep’s life.
  • Calibration is performed manually at one or two points with a trimmer located on the side of the tester.
  • Renewable junction
  • Easy two-point manual calibration
  • 700 hour battery life
  • Simple to use
  • Economical
Specifications
Specifications
±0.1 pH
1.5 V
0.1 pH
95 g
pH Tester
0 to 14
0°C to 50°C
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ pHep™ Instruments pH Tester > pH Tester