Learn More
uvex Pheos B-WR Safety Helmet
Product Code. p-7224529 Shop All Uvex Products
Click to view available options
Color:
White
Unit Size:
Pack of 25
Product Code. 17504116
Brand: uvex 9772030
Only null left
Add to basket
Add to basket
Lightweight safety helmets that feature a sporty design and offer maximum wearer comfort.
  • Lightweight helmet in a sporty design
  • Three variable air vents, one ventilation zone on the forehead and two at the back, for perfect air circulation
  • Adapter systems for uvex visors, uvex earmuffs, and helmet lamps
  • 6-point textile suspension harness with rotating size adjusters for individual with adjustment
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 397
Color White
For Use With (Equipment) Hard Hats
Material HDPE
Model 9772
Product Type Industrial Safety Helmet
Size 52 to 61 cm
Size (Metric) 52 to 61 cm
Style Long Brim
Ventilated Ventilated
Head Style Front Brim