Learn More
Christeyns™ Phago'Derm Sensitive SPS Soap
Click to view available options
Volume (Metric):
500 mL
1000 mL
Quantity:
12 Airless Mounted Pump Bottles
12 Bottles
20 Square Mounted Pump Bottles
Unit Size:
12 L
Pack of 20
Description
- Does not dry out the skin, even with frequent use
- Hypoallergenic (formulated to minimize allergic reactions)
- Suitable for washing children under 36 months and pregnant/breastfeeding women
- Suitable for patient washing and preoperative showering
- Good cleaning power
- High rinsability
- Hydrating potential for 30 min.
Specifications
Specifications
| Product Type | Hand Soap |
| Color | Colorless |
| Form | Foam |
| Quantity | 12 Bottles |
| Volume (Metric) | 1000 mL |