Learn More
SI Analytics™ pH Combination Micro-Electrode
Shop All SI Analytics Products
Click to view available options
Connector Types:
BNC
DIN
DIN, 4 mm Banana Plug
Plug Head
Type:
Combination pH Micro Electrode
pH Combination Electrode
Membrane:
Cylinder
Spear
Sphere
Diameter (Metric):
12/3 mm
3 mm
5 mm
Förpackningsstorlek
Each
Beskrivning
- Proven high-end electrodes for demanding measurement tasks
- Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Each pH combination electrode comes with individual serial number and quality certificate
Specifikationer
Specifikationer
| Connector Types | Plug Head |
| Junction | Ceramic Diaphragm |
| Material | A-Glass |
| pH Range | 7.0 ±0.3 |
| Type | Combination pH Micro Electrode |
| Internal Reference | Ag/AgCl |
| Membrane | Cylinder |
| Temperature (Metric) Range | -5°C to +100°C |
| Length (Metric) | 180 mm |
| Length (Metric) Cable | 1 m |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering