Learn More
pH Buffer Solution pH 5, Fisher Chemical™
Shop All Reagecon Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
10 L
missing translation for 'unitSize'
1 L
10 L
Description
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Specifications
Specifications
| Color | Undesignated |
| pH | 5 |
| Quantity | 1 L |