missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Sachets, METTLER TOLEDO™
GREENER_CHOICE

Product Code. 10457811 Shop All Mettler Toledo Products
Click to view available options
Quantity:
30 x 20 mL
pH:
10.0
4, 7, 10
Unit Size:
597mL
600mL
Each
This item is not returnable. View return policy

Product Code. 10457811

Brand: Mettler Toledo 51302080

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Buffer solutions in sealed sachets for easy and precise calibration of pH measurement equipment.

  • Traceable to NIST primary standards
TRUSTED_SUSTAINABILITY

Specifications

Color Multicolored
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 4, 7, 10
Physical Form Powder or Solid
Quantity 30 x 20 mL
Solution Type pH Buffer
Packaging Satchets
Grade Technical
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.