Learn More
pH 3.776 DIN 19266 Buffer Solution at 25°C, Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Description | Accuracy ± 0.010 pH units |
| pH | 3.76 ±0.010 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | HDPE Bottle GL45 |