Learn More
Masterflex™ PFA Threaded Adapter
Quick and easy assembly
Brand: Masterflex™ 06383-90
Additional Details : Weight : 5.00000kg
Description
- Available in a variety of materials and configurations to meet a wide range of laboratory needs
- Adapts from a NPT male thread to a hose barb.
Specifications
Straight Barbed to Male NPT Threaded Adapters | |
Autoclavable | |
5/32 in. | |
5/32 in. | |
Straight |
Barb x NPT(M) Connection | |
5/32 in. | |
5/32 in. | |
Autoclave or Ethylene Oxide |