Learn More
IKA™ PETTE MicroKit A
GREENER_CHOICE
The IKA PETTE MicroKit A is a starter-friendly pipette package, perfect for any molecular laboratory dealing with low volumes and life science research.
Brand:  IKA™ 0010007638
402.34 EUR valid until 2024-12-31
Use promo code "21904" to get your promotional price.
Product Code. 17513606
  • € 693.00 / Each
In Stock
to see stock.
Explore more special offers
Description
  • The package consists of three single channel pipettes with variable volumes in the micro-volume range, a complimentary stand that holds up to three pipettes and two pipette tip boxes. This makes it a convenient starter package for researchers.
  • Features:
    • Mechanical air-cushion pipettes manufactured in Germany
    • Fully automated calibration by robots
    • Volume-locking feature to prevent any accidental volume changes
    • Large and clearly visible display
    • Ergonomic and lightweight pipette with low pipetting and tip ejection forces to reduce strain and fatigue
    • Durable and robust construction using high quality UV resistant and chemical resistant materials
    • Fully autoclavable to ensure sterility and compliance to stringent laboratory standards
    • Maximum user convenience with tool-less disassembly and calibration adjustment for regular maintenance
Specifications
Specifications
Air Displacement Piston-Operated Pipette
1
Sterile
Variable
Autoclavable
PETTE
Pipette Package
Three Single Channel Pipettes With Variable Volumes In The Micro-Volume Range, One Complimentary Stand That Holds Up To Three Pipettes, And Two Pipette Tip Boxes