Learn More
Brady™ PermaSleeve™ Heat-shrink Labels
Permanently mark wires with the durable PermaSleeve™ PS Heat-shrink polyolefin sleeve label material (B-342). Designed for use with a durable Brady™ handheld label printer.
Brand:  Brady™ 170721
 View more versions of this product
Product Code. 17930760
  • € 61.35 / Pack of 80
In Stock
to see stock.
Explore more special offers
Description
  • Designed specifically for rigorous cable and wire marking applications
  • Self-extinguishing and resistant to extreme temperatures, UV light, chemicals, fuel and oil
  • Fast shrinking to save installation time and cost
  • Made with a crisp, clear identification surface that is resistant to abrasions and fading
Specifications
Specifications
UL Recognised to UL224;SAE-AMS-DTL-23053/5 C (Class 1);SAE AS-81531 Compliant with Directive 2011/65/EU and 2015/863
Non-adhesive
Matte
Polyolefin
Non-sterile
BMP41;BMP51;BMP53;M511;M410;M510
-67°F to 275°F
22 AWG - 16 AWG
0.235 in.
19.05 mm
5.97 mm
M4-125-075-342
White
Die-cut
Wire and Cable Markers
Wire and cable labelling
80
-55°C to 135°C
19.05 mm (W) x 5.97 mm (H)
0.75 in
0.235 in
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ PermaSleeve™ Heat-shrink Labels > Dimensions: 19.05 mm x 5.97 mm; Printer Compatibility: BMP41;BMP51;BMP53;M511;M410;M510; Color: Black on White