Learn More
Brady™ Permanent Paint
Click to view available options
Color:
Black
Blue
Gray
Green
Orange
Red
White
Yellow
Unit Size:
1 pack
Description
- Floor marking paint.
Specifications
Specifications
| Quantity | 6 Pk. |
| Alternate Part Number | EASYLINE PERM. PAINT WHITE (6PC) |
| Product Type | Paint |
| For Use With (Application) | Floor marking |
| Color | White |