Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Brand: Macherey-Nagel™ 723341.30
This item is not returnable.
View return policy
Specifications
0.5 mm | |
0.32 mm | |
G25, G35 | |
220°C to 240°C | |
30 m | |
1 Each |
GC Capillary Column | |
Permabond™ FFAP | |
Gas Chromatography (GC), Free Carboxylic Acids | |
0.25 μm | |
Permabond™ FFAP |
Product Content Correction
Product Title
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column > Length: 30m; I.D.: 0.32mm; Film Thickness: 0.25um