Learn More
Thermo Scientific Chemicals Perindopril tert-butylamine salt
Product Code. 16681852 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 16681852
Brand: Thermo Scientific Chemicals 463210010
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Molecular Weight (g/mol) 441.61
Product Type Perindopril tert-butylamine salt
Quantity 1 g