Learn More
Thermo Scientific™ Oxoid™ Perfringens agar base (tsc/sfp) Oxoid
Brand: Thermo Scientific™ Oxoid™ BO0634M
71.25 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4504305 | Certificate of Analysis | 21/04/2025 | BO0634M |
4502001 | Certificate of Analysis | 07/04/2025 | BO0634M |
4500585 | Certificate of Analysis | 31/03/2025 | BO0634M |
4496236 | Certificate of Analysis | 14/03/2025 | BO0634M |
4496235 | Certificate of Analysis | 11/03/2025 | BO0634M |
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Perfringens agar base (tsc/sfp) Oxoid >