Learn More
BINDER™ Shelf, Perforated
Shop All BINDER Products
Click to view available options
Dimensions (L x W x H):
620 x 510 x 50 mm
Unit Size:
Each
Description
- Shelf
- Perforated
- Made of stainless steel
Specifications
Specifications
| Description | Shelf, perforated - Stainless steel, divided, for divided inner door |
| For Use With (Equipment) | BINDER Chambers CB260, CB-S260 |
| Material | Stainless Steel |
| Type | Divided Shelf |
| Model | CB 260 (E7) |
| Dimensions (L x W x H) | 620 x 510 x 50 mm |
| Length (English) | 2.03 ft. |
| Length (Metric) | 620 mm |
| Width (English) | 1.67 ft. |
| Width (Metric) | 510 mm |
| Show More |