missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctyl iodide, 98%

Product Code. p-180005 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10021721

Brand: Thermo Scientific Acros 269960010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 507-63-1 | C8F17I | 545.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-63-1
Molecular Formula C8F17I
Molecular Weight (g/mol) 545.96
MDL Number MFCD00001064
InChI Key KWXGJTSJUKTDQU-UHFFFAOYSA-N
Synonym perfluorooctyl iodide, heptadecafluoro-1-iodooctane, 1-iodoperfluorooctane, perfluoro-n-octyl iodide, 1-iodoheptadecafluorooctane, perfluoro-1-iodooctane, perfluorooctyliodide, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo
PubChem CID 10491
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
SMILES C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 25°C
Color Colorless to Yellow or Pink
Density 2.0400g/mL
Boiling Point 160°C to 161°C
Flash Point >115°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)7I
Refractive Index 1.3286 to 1.3306
Quantity 1 g
Specific Gravity 2.04
Solubility Information Solubility in water: insoluble
Formula Weight 545.96
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Perfluorooctyl iodide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Perfluorooctyl iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.