Learn More
Pentane, ≥99.0%, Honeywell Riedel-de Haen™
Product Code. 15607050
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Poly Bottle
Unit Size:
1 L
2.5 L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15607050
Brand: Honeywell-Chromasolv 768692.5L
Request Bulk or Custom Format
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CHROMASOLV™, for pesticide residue analysis,≥99.0%
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
Specifications
pH 5.7 (at 22°C)
Melting Point 750°C
Density 0.626g/mL at 25°C
Boiling Point 1393°C (1013 hPa)
Quantity 2.5 L
Explosion Limit 8.3%
Ignition Point 500°C
Packaging Poly Bottle
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Solubility Information 946g/L at 25°C
Formula Weight 72.15g/mol
Percent Purity ≥99.0%
Chemical Name or Material Pentane