Learn More
3M™ PELTOR™ WS™ LiteCom Plus Headset PMR446 MHz
Comes on a headband and features an integrated pre-programmed analog two-way radio, Bluetooth™ Multipoint connectivity, noise-cancelling microphone, and a level-dependent function for ambient listening.
Brand: 3M™ 7100204388
1058.33 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Hearing protector with built-in analogue two-way radio
- Level-dependent technology helps improve situational awareness and communication
- Bluetooth™ multipoint connectivity allows seamless connection of mobile devices
- Push-to-listen quick activation of level dependent function
- Noise cancelling waterproof microphone
- Voice operated transmission (VOX) for hands free use in high noise environments
- Lithium-Ion rechargeable battery
- PMR446 MHz Compatible
- Use to help you communicate efficiently in high noise environments
Specifications
Analog | |
Headband | |
MT73H7A4410WS6EU |