Learn More
3M™ PELTOR™ Battery Charger
3M™ PELTOR™ Battery Charger is designed to be used with 3M™ PELTOR™ ACK03 rechargeable battery pack.
Brand: 3M™ 7000107916
This item is not returnable.
View return policy
Description
- 230 V NiMH Battery Charger
- For use with 3M™ PELTOR™ ACK03 rechargeable battery pack
- Supplied with EU/UK plug adapters
Specifications
Battery Charger | |
PELTOR™ ACK03 Rechargeable Battery Pack | |
EU, UK Plug Adapter |
Product Content Correction
Product Title
3M™ PELTOR™ Battery Charger > Type: Battery Pack