missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Pehanon™ pH Paper

Product Code. 10114954 Shop All Macherey-Nagel Products
Click to view available options
pH Range:
1 to 12
1 to 2.8
10.5 to 13
2.8 to 4.6
3.8 to 5.5
4 to 9
5.2 to 6.8
6 to 8.1
7.2 to 8.8
8 to 9.7
9.5 to 12
Unit Size:
Each
Pack of 200
This item is not returnable. View return policy

Product Code. 10114954

Brand: Macherey-Nagel™ 90401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Measure pH in colored solutions. Pehanon™ pH paper puts the indicator and reference on one strip making it perfect for pH determinations in colored solutions.

PEHANON™ is a special pH-test strips that unifies the pH-indicator and the reference colors on one strip and has the following advantages:

Any sample color has the same effect on both, the reference colors and the reactive pad. This ensures unadulterated readings in colored solutions. The user can be sure to get accurate pH-values.

An invisible hydrophobic barrier just above the top color field prevents capillary action of the test solution beyond. The handle will remain dry and clean whatever the sample is. Therefore the use of the strip is very safe.

pH-values can be determined without a separate color chart. Workers and machinists can use single strips instead of complete packs with color chart which makes PEHANON™ highly economic.

Specifications

pH Range 1 to 12
Format Stick
For Use With (Application) Determination of pH in the colored solutions
Graduations 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12
Quantity 200 Pack
Type pH Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.