Learn More
Trajan Scientific and Medical PEEK Tubing, 360μm O.D., 10 ft
Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs
Brand:  Trajan Scientific and Medical 1304003006-10F
 View more versions of this product
Product Code. 15701059
  • € 69.30 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Description
  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs
Specifications
Specifications
Black
75 μm (0.003 in.)
10 ft.
5000 psi (345 bar)
2 to 14
360 μm
PEEK
100°C