Learn More
asecos™ PE Sump Pallet with Galvanised Grid
Product Code. 17876446 Shop All asecos Products
Click to view available options
Capacity (Metric):
220 L
440 L
520 L
Depth (Metric):
900.00 mm
930.00 mm
1300.00 mm
1330.00 mm
Unit Size:
Each
Product Code. 17876446
Brand: asecos™ 39195
This item requires a price update prior to being ordered.
Request a Price for this item here.
The PE Sump Pallet is approved for the safe storage of water pollutants.
  • PE blue
  • Grid: galvanised
Specifications
Color Blue
Capacity (English) 137.4 gal.
Capacity (Metric) 520 L
Material Polyethylene
Type Painted Spill Sump
Length (English) 99.21 in.
Length (Metric) 2520.00 mm
Width (English) 99.21 in.
Width (Metric) 2520.00 mm
Height (English) 13.78 in.
Height (Metric) 350.00 mm
Depth (English) 35.43 in.
Depth (Metric) 900.00 mm
Weight (English) 163.14 lb.
Weight (Metric) 74 kg