Learn More
Fisherbrand™ PE Plug for Shell Vial
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Outer:
8 mm
12 mm
15 mm
Unit Size:
Pack of 100
Pack of 1000
Description
- PE Plug
- Transparent
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Specifications
| Color | Transparent |
| Diameter (Metric) Outer | 12 mm |
| Material | Polyethylene |
| Quantity | 100 |
| Type | Plug |
| Closure Size | ND12 |