Learn More
asecos™ PE IBC Station with Filling Area and Galvanised Grid
Product Code. 17826346 Shop All asecos Products
Click to view available options
Width (Metric):
1280.00 mm
2460.00 mm
3640.00 mm
Unit Size:
Each
Product Code. 17826346
Brand: asecos™ 40054
This item requires a price update prior to being ordered.
Request a Price for this item here.
The PE IBC Station is approved for the safe storage of water pollutants.
  • Highly resistant against oil, acid, alkali, etc.
  • Easy and safe in-house transport of the empty sump by forklift truck
Specifications
Color Blue
Capacity (English) 264.2 gal.
Capacity (Metric) 1000 L
Material Polyethylene
Type IBC Spill Pallet
Length (English) 143.31 in.
Length (Metric) 3640.00 mm
Width (English) 143.31 in.
Width (Metric) 3640.00 mm
Height (English) 17.72 in.
Height (Metric) 450.00 mm
Depth (English) 70.87 in.
Depth (Metric) 1800.00 mm
Weight (English) 529.10 lb.
Weight (Metric) 240 kg