Learn More
3M™ Particulate Respirator, K100 Series
Click to view available options
Filter Type:
FFP1
FFP2
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 10
Pack of 20
Pack of 200
Description
Provides comfortable, solid, and professional protection at an affordable price.
Specifications
Specifications
| Certifications/Compliance | EN149:2001+A1 2009 NR D |
| Color | White |
| Design Type | Diamond-shaped front, single loop strap |
| Filter Type | FFP1 |
| With Exhalation Valve | Yes |
| Filter Efficiency | FFP1 |
| For Use With (Application) | Exposure to dust particles and /or solid and non-volatile liquid particles |
| Includes | Includes yellow coloured noseclip for easy identification (FFP1) |
| Shape | Diamond shape front |