missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Particle Certified HDPE Containers

Product Code. 18739469
Click to view available options
Capacity (Metric):
125 mL
1000 mL
Description:
HDPE Narrow mouth Nalgene bottle with closure
HDPE Narrow mouth Nalgene bottle with closure, Low Metals
Closure Size:
24-415 mm
38-430 mm
No. per Case:
12
24
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.

Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.

A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.

Key Features:
  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation

Recommended for:

Low particle applications 

 

TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Translucent
Description HDPE Narrow mouth Nalgene bottle with closure, Low Metals
Diameter (English) 3.625 in.
Material HDPE
No. per Case 12
Volume (English) Brim 32 oz.
Volume (Metric) Brim 1000mL
Certifications/Compliance Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Mouth Narrow
Shape Round
Height (Metric) 215.9 mm
Height (English) 8.5 in.
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 92.08 mm
Show More Show Less

†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'