Learn More
Thermo Scientific™ Particle Certified HDPE Containers Product Code.: 17872395
Add to basket
Thermo Scientific™ Particle Certified HDPE Containers
Product Code. 17872395
125 mL, Low Particulate
Click to view available options
Capacity (Metric):
125 mL
1000 mL
Certified:
Low Particulate
Low Particulate, Low Metals
Product Code. 17872395
Brand: Thermo Scientific™ 156125W/N/LP
Add to basket
Add to basket
Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.
Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.
A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.
Key Features:
  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation
Recommended for:
Low particle applications
TRUSTED_SUSTAINABILITY
Specifications
Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Translucent
Diameter (English) 2 in.
Material HDPE
No. per Case 24
Product Line Nalgene
Certified Low Particulate
Volume (English) Brim 4 oz.
Volume (Metric) Brim 125 mL
Certifications/Compliance Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Closure Included Yes
Mouth Narrow
Shape Round
Height (Metric) 101.6 mm
Product Type Bottle
Height (English) 4 in.
Closure Material Polypropylene
Closure Size 24-415 mm
Diameter (Metric) 50.8 mm
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberLP4268010 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number156-125W/N/LP
Lot NumberLP4103100 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog Number156-125W/N/LP
Lot NumberLP3166020 Certificate TypeCertificate of Analysis Date07/07/2023 Catalog Number156-125W/N/LP
Lot NumberLP3145020 Certificate TypeCertificate of Analysis Date07/06/2023 Catalog Number156-125W/N/LP
Lot NumberLP3093040 Certificate TypeCertificate of Analysis Date11/05/2023 Catalog Number156-125W/N/LP
†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Product Content Correction
Product Title
Thermo Scientific™ Particle Certified HDPE Containers >