missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancuronium bromide, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
50 mg
100 mg
0.25 g
Unit Size:
100mg
10mg
250mg
50mg
Specifications
Specifications
| Color | White |
| Quantity | 0.25 g |
| Formula Weight | 732.7 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pancuronium bromide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction