missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium for AAS 1g/L HNO3 matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
€ 169.00 - € 349.00
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | HNO3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12963285
|
Fisher Chemical
B 142 100 |
100 mL |
€ 169.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12973285
|
Fisher Chemical
B 142 500 |
500 mL |
€ 349.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
1g/L | |
HNO3 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for AAS. Palladium (Pd) 1000mg/l in HNO3 5%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .