Learn More
Buerkle™ Packaging Buckets
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
5 L
10 L
Diameter (Metric)Top:
132 mm
175 mm
225 mm
268 mm
Height (Metric) With Closure:
123 mm
131 mm
132 mm
194 mm
263 mm
Unit Size:
Each
Description
Suitable for food or building material industries and non-aggressive chemicals in environmental protection, hosptials and household. Features strong handle and quality closure for product safety.
- White polypropylene
- Air-tight closure
- Includes lid
Specifications
Specifications
| Product Type | Packaging Bucket |
| Capacity (Metric) | 3 L |
| Material | Polypropylene |
| Color | White |
| Diameter (Metric)Top | 225 mm |
| For Use With (Equipment) | Suitable for The Food or Building Material Industries and Non-Aggressive Chemicals in Environmental Protection, Hospitals and The Household |
| Height (Metric) With Closure | 123 mm |
| Includes | Quality Closure and Strong Lid |
| Quantity | 1 Ea. |