Learn More
Thermo Scientific™ Oxoid™ Nalidixic Acid Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Brand:  Thermo Scientific™ CT0031B
44.76 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10249712
  • € 74.80 / Pack of 250
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Nalidixic Acid Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nalidixic Acid
30µg
NA
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with CLSI, BSAC, EUCAST, SFM guidelines
Oxoid Nalidixic Acid Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6243695 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberCT0031B
Lot Number6227130 Certificate TypeCertificate of Analysis Date19/02/2025 Catalog NumberCT0031B
Lot Number6206018 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog NumberCT0031B
Lot Number6173323 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberCT0031B
Lot Number6196708 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberCT0031B
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Nalidixic Acid Antimicrobial Susceptibility discs > 30μg