Learn More
Thermo Scientific™ Oxoid™ Compact Plastic Pouch
Thermo Scientific™ Oxoid Compact Plastic Pouch is for use with Compact System.
Brand:  Thermo Scientific™ AG0020C
52.67 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10508263
  • € 67.00 / Pack of 20
In Stock
to see stock.
Explore more special offers
Description
Thermo Scientific™ Oxoid Compact Plastic Pouch is for use with Compact System.
  • Use with AnaeroGen™ Compact Sealing Clip Part No. AN0005C
  • AnaeroGen Compact sachet, Part No. AN0020C, CO22Gen Compact sachet Part No. CN0020C, and sachet Part No. CD0020C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Compact Plastic Pouch
Compact Pouch
Thermo Scientific Oxoid
20/Pk.
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6251118 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog NumberAG0020C
Lot Number6229818 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog NumberAG0020C
Lot Number6220668 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberAG0020C
Lot Number6215242 Certificate TypeCertificate of Analysis Date25/12/2024 Catalog NumberAG0020C
Lot Number6208731 Certificate TypeCertificate of Analysis Date14/12/2024 Catalog NumberAG0020C
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Compact Plastic Pouch > Pack of 20