missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ Oxivir CE Plus Spray
Cleaner and Disinfectant for non-invasive medical devices
Brand: Diversey™ 100829237
92.70 EUR valid until 2024-09-30
Use promo code "21675" to get your promotional price.
This item is not returnable.
View return policy
Description
- Patented Hydrogen Peroxide based acidic formulation
- Short contact times
- Effective Cleaner
- Fragrance free
- Extensively tested against European norms and effective against multiple pathogens of concern, including Coronavirus, HIV, HBV, HCV, Avian Influenza, Pseudomonas aeruginosa , Staphylococcus aureus, Salmonella enterica, MRSA, VRE and Norovirus
- Cleans and disinfects in one step, reducing working time
- Excellent cleaning performance, including descaling and lime limitation removes the need for a pre-cleaning step
- Good user safety profile, with no hazard warnings on RTU format; does not contain respiratory irritants
- AHP formulation is gentle on surfaces extending equipment life
- Good environmental profile. Active breaks down into oxygen and water
- Suitable for use in high risk areas in Healthcare, including OR, ICU, NICU and patient rooms
- Medical Device Class IIa
Specifications
Disinfectant Spray | |
Non-sterile | |
Spray Bottle | |
6 Pk. |
1.8 | |
Patented Hydrogen Peroxide based acidic formulation | |
For BE, NL, PT only | |
750 mL |