Learn More
WTW™ OxiTop™ Measuring Vessel
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
Brand:  WTW™ 209019
197.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10528401
  • € 208.00 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Specifications
1000 mL
OxiTop™-C Accessories
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WTW™ OxiTop™ Measuring Vessel > Capacity: 1000mL