Learn More
Haldenwanger™ Oxide Ceramic Alsint Conical Flat Bottom Crucible
Product Code. 10058480
Click to view available options
Capacity (Metric):
270 mL
Unit Size:
Each
Product Code. 10058480
Brand: Haldenwanger™ L ALSINTTIEGEL*A/6
Only null left
Add to basket
Add to basket
Order Info
CapacityMetric
Specifications
Material Oxide Ceramic
Capacity (Metric) 270 mL
Diameter (Metric) Outer 65mm
Product Type Alsint Crucible
Shape Cylinder