Learn More
Thermo Scientific™ Owl™ Sequencing System Spacer Sets and Glass Plates
Shop All Thermo Scientific Products
Click to view available options
Product Type:
Offset Glass
Height (Metric):
0.18 in.
Unit Size:
Each
Description
Use Thermo Scientific™ Owl™ Sequencing System Spacer Sets and Glass Plates with Owl™ Sequencing Gel Casting Systems.
Specifications
Specifications
| Description | Offset Glass; 43cmL x 35cmW x 0.18 in. thickness |
| For Use With (Equipment) | Owl™ Sequencing System |
| Height (Metric) | 0.18 in. |
| Length (Metric) | 43 cm |
| Product Type | Offset Glass |
| Thickness (Metric) Teeth | 3/16 in. |
| Width (Metric) | 35 cm |