missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts

Product Code. p-4528800 Shop All Thermo Scientific Products
Click to view available options
Type:
Buffer Chamber and Lid
Casting Slides
Gel Casting Tray
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12474577

Brand: Thermo Scientific™ C2

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts.

Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts. Parts are for use with the Owl C2-S Micro Electrophoresis System, a system designed for quick restriction fragment analysis.

  • Glass slides, 5 x 7.5 cm, are used as the gel tray
  • Thin gels can be poured and formed by surface tension directly on the glass slide
TRUSTED_SUSTAINABILITY

Specifications

Capacity 100 mL
For Use With (Equipment) Owl™ C2-S Micro Electrophoresis System
Product Line Owl
Type Buffer Chamber and Lid
Length (Metric) 17 cm
Width (Metric) 6.5 cm
Height (Metric) 6.5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.