Learn More
Thermo Scientific™ Owl™ Accessories for CST Multiple Gradient Gel Casters
Shop All Thermo Scientific Products
Click to view available options
Type:
Binder Clamp
Clamp
Unit Size:
Each
Description
Choose between stainless-steel clamps or binder clamps from Thermo Scientific™ Owl™ Accessories for CST Multiple Gradient Gel Casters. Packaged in sets of 12, these clamps are for use with Owl CST Multiple Gradient Gel Casters.
Specifications
Specifications
| For Use With (Equipment) | Owl CST System |
| Product Line | Owl |
| Type | Clamp |
| Material | Stainless Steel |
| Quantity | 12 Pk. |