Learn More
Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Standard Comb
Product Code. 12483647 Shop All Thermo Scientific Products
Click to view available options
Type:
Rapid Load Comb
Teeth Count:
25 (2X)
9 (1X)
Thickness (Metric) Teeth:
1.0 mm
1.5 mm
Unit Size:
Each
Product Code. 12483647
Brand: Thermo Scientific™ A1RL9C
Only null left
Add to basket
Add to basket
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs. For use with the Fisherbrand™ Owl™ A1 Large Gel Electrophoresis Systems.
Eliminate comb assembly with this assortment of heavy-duty, one-piece Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Combs, for use with Owl A1 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl A1 Large Gel Electrophoresis System
Product Line Owl
Type Rapid Load Comb
Teeth Count 9 (1X)
Thickness (Metric) Teeth 1.0 mm