Learn More
Honeywell™ Overspec Safety Glasses
Product Code. 12637423
Clear, Medium
Click to view available options
Lens Tint:
Clear
Gray
Size:
Medium
Large
OTG protection featuring exceptional peripheral vision/coverage, and a customizable fit.
  • Over-the-glass coverage, allowing workers full protection while wearing prescription eyewear
  • Adjustable and bendable
  • Wire-core temples offer flexibility for a secure, customized fit
  • Ratchetting temples (3 positions) to further customize fit
  • For use in agriculture, manufacturing, and pharmaceutical industries
  • Protection from abrasion, airborne particulates, biohazard, impact
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: C899.1B-D, EC Attestation: C899.3B-D
Lens Coating Anti-fog
Lens Tint Clear
For Use With (Application) For use in agriculture, manufacturing, and pharmaceutical industries
Frame Color Blue, Black
Size Medium
Lens Material Polycarbonate
Frame Material Nylon
Product Content Correction
Product Title
Honeywell™ Overspec Safety Glasses >