Learn More
3M™ Overflow Valves
Click to view available options
Color:
Black
Unit Size:
1 pair
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Versaflo Supplied Air System S-200 |
| Model | C-340 |
| Quantity | 2/Pk. |
| Product Type | Overpressure Valve |