Learn More
IKA™ Oven 125 Control Dry Glass
Shop All IKA Products
Click to view available options
Capacity (Metric):
125 L
Unit Size:
Each
Description
- Rapid heating and exact temperature regulation guarantee reproducible results in industrial laboratories, schools and universities, research, quality control or science. Thanks to its high quality insulation, the device has a very large interior of 125 L in relation to its total volume. This structure also reduces odour emissions and operating costs and ensures excellent temperature uniformity.
- The Oven 125 control dry recirculating air drying cabinet heats up to 300°C. The additional temperature selection limited TWB (DIN 12880) meets the highest security requirements. Furthermore, the temperature curve can be monitored during testing independently of the safety temperature. This means that your test material is always protected.
- The glass door allows for better monitoring of your samples.
- The generously dimensioned interior of the Oven 125 control dry is comfortably illuminated. A practical cable bushing at the rear of the device is provided as standard for measurements in the oven or in the test material. The fan switches off automatically if the door is open. Up to 6 insertable shelves can be accommodated in the interior made of stainless steel.
- The Oven 125 control dry offers an interface for an additional sensor for direct temperature regulation in the test material. Test parameters can be stored directly on a USB stick.
- All important process parameters are displayed clearly on two large TFT displays. The temperature profiles and individual user profiles can be entered via the intuitive user interface. The real-time clock enables convenient digital documentation and a time-delayed start.
Specifications
Specifications
| Capacity (Metric) | 125 L |
| Temperature (Metric) | Ambient to 300°C |
| Display | LCD |
| Max. Shelf Load | 30 kg |
| No. of Shelves | 6 |
| Timer | 1 to 142560 min |
| Dimensions (D x W x H) Interior | 450 x 550 x 525 mm |
| Dimensions (L x W x H) | 650 x 700 x 825 mm |
| Weight (Metric) | 82 kg |
| Voltage | 100 to 115 V, 220 to 240 V |
| Show More |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur