Learn More
ORP Test Solution, 200 to 275 mV, Hanna Instruments™
Click to view available options
Quantity:
230 mL
Specifications
Specifications
Quantity | 230 mL |
Odor | Odorless |
Physical Form | Liquid with Precipitate |
Concentration | 200 to 275 mV |
Color | Brown/Orange |
pH | 4 |
Solubility | Soluble |
Product Content Correction
Product Title
ORP Test Solution, 200 to 275 mV, Hanna Instruments™ >