Learn More
Thermo Scientific™ Orion™ Ionplus™ Sure-Flow™ Lead Combination Ion Selective Electrode
Click to view available options
Connector Types:
BNC
BNC (Waterproof)
Screw Cap
Unit Size:
Each
Description
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
- Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials
- When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap
- Lead Half-Cell Electrode used in conjunction with the Orion 900200 Double Junction Reference electrode offers the benefits of the Sure-Flow™ junction design
- Calibration standards catalog number: 948206 - 0.1 M Pb(CIO4)2; 948207 - 0.1 M Na2SO4
- Required ISA Catalog numbers: Consult instruction manual
Specifications
Specifications
| Connector Types | Screw Cap |
| Description | With screw cap connector |
| Drift | drift-free potentials |
| Junction | Sure-Flow Junction |
| For Use With (Equipment) | ISE Meter |
| Type | Lead Ion Selective Electrode, screw cap connector |