missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ISE Calibration Standards

Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.

Brand:  Thermo Scientific™ 920706

 View more versions of this product

Product Code. 12750588

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.

All standards are 1 pint (475mL) bottles, unless otherwise noted.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitrate Standard
475 mL
C of A Acceptance Range: 0.100 M +/- 0.002 M Potassium Nitrate
Nitrate Ion Selective Electrodes Including 9707BNWP, 9307BNWP
0.1 M NaNO3
Nitrate Ion Selective Electrode
Calibration Standard
1 Bottle
Yes
Yes
Liquid
Colorless
Calibration Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCO1 Certificate TypeCertificate of Analysis Date30/12/2024 Catalog Number920706
Lot NumberCR1 Certificate TypeCertificate of Analysis Date30/12/2024 Catalog Number920706
Lot NumberCS1 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog Number920706
Lot NumberCZ1 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number920706
Lot NumberBR1 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog Number920706
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ ISE Calibration Standards > For Nitrate Ion measurements. 0.1M NaNO3; 475mL; CAS: (7631-99-4)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.